Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydrazinylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-hydrazinylpyridine2-Hydrazinopyridine2(1H)-Pyridinone, hydrazone

12087-0250 12087-0010 12087-0050
TS12087-0010EA 36.95 USD InStock
TS12087-0010 TS12087-0050 TS12087-0250
2-Hydrazinylpyridine 98%
2-Hydrazinylpyridine


Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 185 °C (140 mmHg)
Melting Pt: 44.5…46 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006249
CAS Number: 4930-98-7
Order Now
Specification Test Results
Appearance White to orange-brown Crystals or crystalline mass
Infrared spectrum Conforms
Melting point 43 to 47 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty