Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylimidazole 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methyl-1H-imidazole

12700-0010 12700-5000 12700-1000
TS12700-5000EA 79.93 USD InStock
TS12700-5000 TS12700-0010 TS12700-1000
2-Methylimidazole 99%
2-Methylimidazole


Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 142…145 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 3263
ADR: 8,III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 142 to 145 °C
Water ≤0.5 % (K.F.)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty