Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Piperonyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 3,4-(Methylenedioxy)benzyl alcohol1,3-Benzodioxol-5-ylmethanol

13135-1000 13135-0250
TS13135-1000EA 114 USD InStock
TS13135-1000 TS13135-0250
Piperonyl alcohol 98%
Piperonyl alcohol


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 156…158 °C (66 mmHg)
Melting Pt: 51…55 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005836
CAS Number: 495-76-1
EINECS: 207-808-4
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 51 to 55 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty