Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Quinuclidinone hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride

13226-5000 13226-1000 13226-0250
TS13226-5000EA 746 USD InStock
TS13226-5000 TS13226-0250 TS13226-1000
3-Quinuclidinone hydrochloride 99%
3-Quinuclidinone hydrochloride


Formula: C₇H₁₂ClNO
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00137391
CAS Number: 1193-65-3
EINECS: 214-776-5
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration Argentometric 98.5 to 101.5 % (On dry substance)
Loss on drying ≤1.0 % (100°C, 2 h)
Heavy metals ≤10 ppm
Sulfated ash ≤0.1 % (800°C)
UV (6 M in H2O)
UV at 260 nm A: ≤0.06
UV at 280 nm A: ≤0.06
pH 6.0 to 9.5 (20°C, 10% in H2O)
Titration with HClO4 99.0 to 101.0 %
Chloride (Cl) ≤0.05 %
Sulfate (SO4) ≤0.05 %
Ammonium (NH4) ≤0.05 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty