Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxetane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,3-Trimethylene OxideTrimethylene oxide

14014-0250
TS14014-0250EA 385 USD InStock
TS14014-0250
Oxetane 97%
Oxetane


Formula: C₃H₆O
Density: 0.89 g/cm³
Flash Pt: <1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005167
CAS Number: 503-30-0
EINECS: 207-964-3
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3920 to 1.3940 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty