Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Pyrazolamine1H-Pyrazol-3-amine3-Aminopyrazole3-AP

17103-0100 17103-0025
TS17103-0100EA 192 USD InStock
TS17103-0100 TS17103-0025
3-Amino-1H-pyrazole 98%
3-Amino-1H-pyrazole


Formula: C₃H₅N₃
MW: 83.09 g/mol
Boiling Pt: 218 °C (122 mmHg)
Melting Pt: 33…39 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005236
CAS Number: 1820-80-0
EINECS: 217-338-1
UN: 3263
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to light brown Liquid or fused solid
Infrared spectrum Conforms
GC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty