Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-4,6-dichloropyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4,6-Dichloropyrimidin-5-ylamine

20325-0250 20325-0050
TS20325-0250EA 569 USD InStock
TS20325-0250 TS20325-0050
5-Amino-4,6-dichloropyrimidine 97%
5-Amino-4,6-dichloropyrimidine


Formula: C₄H₃Cl₂N₃
Melting Pt: 143…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00006108
CAS Number: 5413-85-4
EINECS: 226-503-7
Order Now
Specification Test Results
Appearance Yellow-brown Powder or crystals
Infrared spectrum Conforms
Melting point 143 to 148 °C
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty