Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R)-2-(Aminomethyl)-1-ethylpyrrolidine

35700-0010 35700-0050
TS35700-0050EA 343 USD InStock
TS35700-0050 TS35700-0010
(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine 97%
(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine


Formula: C₇H₁₆N₂
Density: 0.929 g/cm³
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00671482
CAS Number: 22795-97-7
UN: 2734
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4655 to 1.4685 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation +98° to +108° (20°C, 589 nm) (c=1, CH3OH)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty