Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-O-Isopropylidene-D-glyceraldehyde 97%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde(4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde

36926-0010
TS36926-0010EA 131 USD InStock
TS36926-0010
2,3-O-Isopropylidene-D-glyceraldehyde 97%
2,3-O-Isopropylidene-D-glyceraldehyde


Formula: C₆H₁₀O₃
MW: 130.143 g/mol
Boiling Pt: 139 °C (760 mmHg)
Density: 1.045 g/cm³
Flash Pt: 62 °C
Storage Temperature: Freezer
MDL Number: MFCD00269682
CAS Number: 15186-48-8
Order Now
Specification Test Results
Appearance Clear to hazy colorless to yellow Liquid or solid
Infrared spectrum Conforms
Additional info Contains various amounts of polymer
Additional info Crack to monomer by distillation before use
GC ≥96.0 %
Specific optical rotation +39° to +69° (20°C, 589 nm) (C=2, Chloroform)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty