Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-γ-Hexalactone 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (±)-γ-Caprolactone(±)-Hexan-4-olideγ-Caprolactone5-Ethyldihydrofuran-2(3H)-one

39417-0250 39417-5000 39417-1000
TS39417-5000EA 133 USD InStock
TS39417-5000 TS39417-0250 TS39417-1000
(±)-γ-Hexalactone 98%
(±)-γ-Hexalactone


Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: –18 °C
Density: 1.023 g/cm³
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00005401
CAS Number: 695-06-7
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty