Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,6-Dibromocarbazole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,6-Dibromo-9H-carbazole

43397-0050
TS43397-0050EA 76 USD InStock
TS43397-0050
3,6-Dibromocarbazole 97%
3,6-Dibromocarbazole


Formula: C₁₂H₇Br₂N
Melting Pt: 212…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00004961
CAS Number: 6825-20-3
Order Now
Specification Test Results
Appearance Tan Crystalline powder
Infrared spectrum Conforms
Melting point 211 to 215 °C
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty