Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-1-Boc-3-hydroxypiperidine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate(3S)-3-Hydroxypiperidine, N-BOC protected

43644-0050
TS43644-0050EA 374 USD InStock
TS43644-0050
(S)-1-Boc-3-hydroxypiperidine 97%
(S)-1-Boc-3-hydroxypiperidine


Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 48.9…51.7 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115307
CAS Number: 143900-44-1
Order Now
Specification Test Results
Appearance White to tan Powder
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +9° to +11° (20°C, 589 nm) (C=1 in chloroform) / +21° to +24° (20°C, 589 nm) (c=5 in MeOH)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty