Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Thiophenemethanol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Thiophen-3-ylmethanolThiophen-3-methanol3-Thienylmethanol

44224-0050 44224-0250
TS44224-0050EA 96 USD InStock
TS44224-0050 TS44224-0250
3-Thiophenemethanol 97%
3-Thiophenemethanol


Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 73 °C (6Torr)
Density: 1.211 g/cm³
Flash Pt: 100.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014534
CAS Number: 71637-34-8
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty