Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ceftazidime pentahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

46173-0050 46173-0250
TS46173-0250EA 277.5 USD InStock
TS46173-0250 TS46173-0050
Ceftazidime pentahydrate 98%
Ceftazidime pentahydrate


Formula: C₂₂H₃₂N₆O₁₂S₂
Melting Pt: >150 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00153936
CAS Number: 78439-06-2
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water 13 to 15 % (0.1g)
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty