Would you like to log in again?
Whatman™ Grade 595 ½ Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10311642 10311647 10311645 10311654 10311641 10311642 10311653 10311649 10311643 10311656 10311647 10311645 10311654 10311651 10311656 10311643 10311644 10311652 10311652 10311649 10311653 10311641 10311651 10311644
10034-660PK 407.22 USD InStock
10034-660 10034-650 CA11012-760 CA11012-672 10034-646 CA11012-674 CA11012-684 10034-656 10034-648 CA11012-752 10034-658 CA11012-664 CA11012-678 CA11012-666 10034-652 CA11012-702 10034-644 10034-654 CA11012-704 CA11012-758 10035-782 CA11012-708 10035-784 10035-786
Whatman™ Grade 595 ½ Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A prepleated filter used for many routine analytical applications in different industries (e.g., particle separation from food extracts or filtration of solids from digested environmental samples, e.g., for ICP/AAS analysis).
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself
  • The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles.
Grade 595 ½: 4 - 7 µm
Very popular, thin filter paper, medium-fast with medium to fine particle retention. Used for many routine analytical applications in different industries (e.g., particle separation from food extracts or filtration of solids from digested environmental samples, e.g., for ICP/AAS analysis).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty