Would you like to log in again?
United Scientific™ Multichannel Micropipettes, 12 Channel
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
PMC12-200 PMC12-100 PMC12-50 PMC12-300 PMC12-10 PMC12-20
470339-588EA 419 USD InStock
470339-588 470340-114 470338-992 470338-892 470338-956 470339-726
United Scientific™ Multichannel Micropipettes, 12 Channel
Pipettes Micropipettes
Fully autoclavable 12 channel micropipette.
  • Ideal for ELISA, molecular screening, kinetic studies, DNA amplifications and more
  • Designed with click-stop volume control, built-in tip ejectors and comfortable grips to ensure accuracy and maximize lab productivity
These multichannel micropipettes are recommended for a wide variety of applications such as ELISA, molecular screening, kinetic studies, and DNA amplifications. Also available in 8-channel options.
The variable volume micropipettes feature built-in tip ejectors and autoclavable tip cones. They work on a click stop digital system, are easy to calibrate and maintain, and easy to disassemble for autoclaving.
Certifications: Manufactured in an ISO 9001:2008 certified facility, these pipettes are highly accurate. Each pipette is individually calibrated according to ISO 8655 standards and includes a calibration certificate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty