Would you like to log in again?
Test Tube Drying Rack, United Scientific Supplies
Supplier: United Scientific Supplies
77851
470339-148PK 46.1 USD InStock
470339-148
Test Tube Drying Rack, United Scientific Supplies
Racks Tube Racks
Drying racks for tubes and plates.
  • Autoclavable
  • Inverted tubes design to minimize collection of airborne contaminants
These polypropylene racks can conveniently hold inverted tubes for drying and minimize collection or airborne contaminants. Racks can also be used to hold and dry electrophoresis and chromatography plates. Autoclavable. The 96 place rack features an 8×12 array of pegs that hold test tubes up to 13 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty