Would you like to log in again?
Digital Timer with Photogates, United Scientific Supplies
Supplier: United Scientific Supplies
DTPHG1 DTPHG1-P
470339-940EA 326.15 USD InStock
470339-940 470339-974
Digital Timer with Photogates, United Scientific Supplies
Timers Digital Timers
Accurately measure acceleration due to gravity.
  • Four-digit timer designed for use with air tracks, free fall apparatuses, and other classroom equipment
  • Microprocessor-controlled, with eight programmed functions
  • Comes with two photogates
This four-digit timer is designed to be used with air tracks, free fall apparatuses, and other equipment. It is microprocessor-controlled and offers eight programmed functions to record counts, interruption timing, time between interruptions, period of oscillation, acceleration, acceleration due to gravity, velocity in collisions, and time scale output. Includes two photogates and activity guide.
Delivery information: Includes activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty