Would you like to log in again?
Racked Micro Tips, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
P10148 P10117 P10111 P10113
470339-346EA 88.3 USD InStock
470339-346 470339-866 470339-134 470339-428
Racked Micro Tips, Sterile, United Scientific Supplies
Pipette Tips
Convenient rack and sterile micropipette tips.
  • Inner radial rings enable airtight grip on tip cone
  • Manufactured in a precision molding facility from 100% virgin polypropylene
  • DNase, RNase and Pyrogen free
Case of 10 racked, sterile pipette tips. Each rack contains 96 tips, size 2 to 200 μl, for a total of 960 tips.
The highly smooth, hydrophobic surface provides low liquid retention, increased accuracy and repeatability.
Color: Yellow
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty