Would you like to log in again?
Filter Micro Tips, United Scientific Supplies
Supplier: United Scientific Supplies
P11005 P11007
470339-694EA 45.9 USD InStock
470339-694 470340-242
Filter Micro Tips, United Scientific Supplies
Pipette Tips
Filtered pipette tips to prevent cross-contamination.
  • Filters manufactured from 100% pure porous polyethylene
  • Designed to prevent liquids and aerosols from entering the body of the micropipette to protect against cross-contamination
  • Beveled and graduated tips are made from ultrapure USP Class VI certified, virgin polypropylene
  • Non-cytotoxic and non-hemolytic, with a 99.9% bacterial filtration efficiency (BFE)
  • DNase, RNase and Pyrogen free
Pack of 1000 filter pipette tips, size 2 to 200 μl. Manufactured from 100% pure porous polyethylene, filters are designed to prevent liquids and aerosols from entering the body of the micropipette to protect against cross-contamination. Best suited for biological applications, these tips are non-cytotoxic, non-hemolytic, and have a 99.9% bacterial filtration efficiency (BFE). Tips are manufactured from ultrapure USP Class VI certified, virgin polypropylene.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty