Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Wooden Test Tube Rack, 12 Holes, for 28 mm tubes

Supplier: EISCO SCIENTIFIC LLC
CH0005C
470345-678EA 24.9 USD InStock
470345-678
Eisco Wooden Test Tube Rack, 12 Holes, for 28 mm tubes
Racks Tube Racks
Accommodates larger diameter tubes.

  • Made of strong and durable polished beechwood - easy to clean, built to last
  • Holds 12 tubes up to 28 mm upright
  • Can be repurposed to hold other lab utensils, pens, pencils, and markers

Premium quality wooden rack designed to hold up to 12 test tubes vertically. Polished. Rack features 12 holes measuring 28 mm in Ø.

Dimension: 10.25L×3D×6"H

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty