Would you like to log in again?
Eisco Wooden Test Tube Rack, 13-Holes, 6 Pins
Supplier: EISCO SCIENTIFIC LLC
CH0003C
470344-884EA 30.2 USD InStock
470344-884
Eisco Wooden Test Tube Rack, 13-Holes, 6 Pins
Racks Tube Racks
Compact design optimizes lab bench space.
  • Sturdy beech wood construction will stand up to the wear and tear of the lab
  • Holds 13 tubes upright and 6 tubes inverted
  • Can be repurposed to hold other lab utensils, pens, pencils, and markers
Premium quality wooden rack made from polished beach wood. Top shelf of rack has 7 holes that accommodate tubes of up to 20 mm, and bottom has 6 holes that accommodate tubes up to 25 mm. Measures 10.25" wide, 3.25" long and 8" tall.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty