Would you like to log in again?
Centrifuge Tube Boxes, PP, United Scientific Supplies
Supplier: United Scientific Supplies
P20604 P20605
470340-216EA 88.4 USD InStock
470340-216 470339-450
Centrifuge Tube Boxes, PP, United Scientific Supplies
Racks Tube Racks
Durable PP centrifuge tube box.
  • Durable alternative to cardboard or foam tube racks
  • Tube placeholders contain numeric code, easily viewed through included clear lid
  • Autoclavable
Durable polypropylene construction provides a rugged alternative to cardboard or foam racks. Can withstand a temperature range of –90 to 121 °C (–130 to 250 °F). Each tube placeholder contains a numeric code, easily visible through the clear lid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty