Would you like to log in again?
Test Tube Racks, Wire, Epoxy-Coated, United Scientific Supplies
Supplier: United Scientific Supplies
TTWE02 TTWE03 TTWE04 TTWE01
470339-762EA 24.05 USD InStock
470339-762 470339-560 470339-878 470339-804
Test Tube Racks, Wire, Epoxy-Coated, United Scientific Supplies
Racks Tube Racks
Heavy duty epoxy coated wire racks.
  • Heavy wire test tube rack with durable epoxy coating
  • Autoclavable
Available for multiple tube sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty