Would you like to log in again?
Porcelain Desiccator Plates, Large Holes, United Scientific Supplies
Supplier: United Scientific Supplies
JDS230 JDS095 JDS146 JDP230 JDS190 JDP140 JDS115 JDP190 JDS142
470340-126EA 35.7 USD InStock
470340-126 470339-018 470339-672 470339-144 470338-984 470339-266 470339-586 470339-950 470338-908
Porcelain Desiccator Plates, Large Holes, United Scientific Supplies
Desiccators Desiccators, Plates
Porcelain desiccator plates with large holes.
  • Autoclavable
  • Glazed on top surface
  • No glaze softening occurs up to 1000 °C
Plates are glazed on top surface and is resistant to acids and alkalies, excluding hydrofluoric acid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty