Would you like to log in again?
Micropipette Tips, Low Retention, PP, United Scientific Supplies
Supplier: United Scientific Supplies
P10004 P10003 P10005 P10006 P10001 P10002
470339-876EA 36.5 USD InStock
470339-876 470339-426 470339-860 470339-268 470339-758 470339-826
Micropipette Tips, Low Retention, PP, United Scientific Supplies
Pipette Tips
Reduce sample retention for more precision.
  • Highly homogenous surface is free of defects, preventing non-specific binding
  • Manufactured from ultrapure USP Class VI certified, virgin polypropylene
  • DNase, RNase and Pyrogen free
  • Natural color
Deliver 10X lower retention levels with viscous samples, as compared to standard pipette tips.
Packaging: Pack of 1000 graduated micropipette tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty