Would you like to log in again?
Racked Filter Micro Tips, United Scientific Supplies
Supplier: United Scientific Supplies
P11037 P11025 P11029 P11033
470338-972EA 111.4 USD InStock
470338-972 470339-256 470339-704 470339-914
Racked Filter Micro Tips, United Scientific Supplies
Pipette Tips
Conveniently rack and sterile pipette tips.
  • Filters manufactured from 100% pure porous polyethylene
  • Tips made from ultrapure USP Class VI certified, virgin polypropylene
  • DNase, RNase and Pyrogen free
  • Case of 10 racked, sterile filter pipette tips
Each rack contains 96 graduated tips for a total of 960 tips. Designed to prevent liquids and aerosols from entering the body of the micropipette to protect against cross-contamination. Best suited for biological applications, these tips are non-cytotoxic, non-hemolytic, and have a 99.9% bacterial filtration efficiency (BFE).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty