Would you like to log in again?
Pipette Filling Device, United Scientific Supplies
Supplier: United Scientific Supplies
MPF-45-PK/5 MPF-20-PK/5 PFD001 PFDS01
470339-096EA 40.35 USD InStock
470339-096 470339-152 470339-290 470339-606
Pipette Filling Device, United Scientific Supplies
Pipettes Pipette Controllers
Electronic pipette filling device.
  • Electronic device allows for quick and easy filling of pipettes
  • Universal silicon collate accepts standard pipettes up to 100 ml
  • Designed for up to 8 hours of continuous operation
  • Included battery charger plugs into standard wall outlet
Fill any standard pipette using this efficient and reliable electronic instrument. It is designed for ease and comfort to suit various types of liquid handling needs in laboratories. Features include LED charging indicators, adjustable pump speed, comfort grip and multiple housing color combinations. A universal silicon collet is provided to accept all standard pipettes up to 100 ml. Internal knurlings on the collet hold the pipette firmly.
An in-line membrane filter (0.2 µm filter included) prevents aerosol contamination and prevents liquid from entering the equipment by accident.
Ni-MH 3.6 V batteries are used in combination with a low power consumption circuit and vacuum/pressure pump. This allows for eight hours of continuous operation. Each device includes a battery charger that plugs into a standard wall outlet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty