Would you like to log in again?
Support Stands with Rod, Porcelain, United Scientific Supplies
Supplier: United Scientific Supplies
SSP6X6 SSP7X13
470339-342EA 110.25 USD InStock
470339-342 470339-948
Support Stands with Rod, Porcelain, United Scientific Supplies
Support Stands
Porcelain coated support stands.
  • Heavy porcelain base with glazed finish to prevent chemical seepage
  • Removable aluminum support rod
A hole for the removable support rod is conveniently located in the center of the base, and rubber feet are affixed to the base bottom to prevent sliding and/or damage to the work surface.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty