Would you like to log in again?
SureSTART™ Specification Certified 20 ml Screw Clear Glass Vial and 24 mm Closed Top Screw Cap Kits for Scintillation, Level 2, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6AK20SCIN
TS6AK20SCINPK 437.23 USD InStock
TS6AK20SCIN
SureSTART™ Specification Certified 20 ml Screw Clear Glass Vial and 24 mm Closed Top Screw Cap Kits for Scintillation, Level 2, Thermo Scientific
Vials
Perform accurate liquid scintillation counting while safely containing radioactive cocktails with our level 2 Thermo Scientific™ SureSTART™ specification certified screw vial and cap kits for scintillation.
Select level 2 products for high throughput applications, when robustness and reproducibility are key. Our kits for scintillation include 20 ml glass screw vials and closed top polypropylene caps lined with polypropylene foil septa.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty