Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

(±)-2-Chloromandelic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloromandelic acid(±)-2-Chlorophenylglycolic acid

24068-0500 24068-2500 24068-0100
TS24068-0500EA 154 USD InStock
TS24068-0500 TS24068-2500 TS24068-0100
(±)-2-Chloromandelic acid 98%
(±)-2-Chloromandelic acid


Formula: C₈H₇ClO₃
MW: 186.59 g/mol
Melting Pt: 80…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00084962
CAS Number: 10421-85-9
EINECS: 233-900-9
Order Now
Specification Test Results
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 88 to 93 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty