Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylacetyl chloride 97.5%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,2-Diphenylacetyl chloride

29202-0050 29202-0250
TS29202-0250EA 168 USD InStock
TS29202-0250 TS29202-0050
Diphenylacetyl chloride 97.5%
Diphenylacetyl chloride


Formula: C₁₄H₁₁ClO
MW: 230.69 g/mol
Boiling Pt: 175…176 °C (17 mmHg)
Melting Pt: 51…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013655
CAS Number: 1871-76-7
EINECS: 217-493-5
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥96.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty