Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Cyanostyrene 95% stabilized

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Ethenylbenzonitrile

29291-0100 29291-0025
TS29291-0100EA 466 USD InStock
TS29291-0100 TS29291-0025
4-Cyanostyrene 95% stabilized
4-Cyanostyrene


Formula: C₉H₇N
MW: 129.162 g/mol
Boiling Pt: 56…58 °C (10 mmHg)
Storage Temperature: Freezer
MDL Number: MFCD00080445
CAS Number: 3435-51-6
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5765 to 1.5785 (20°C, 589 nm)
GC ≥94 %
Stabilizer 0.05 % tert.butylcatechol

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty