Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-(trifluoromethyl)phenyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-4-isocyanato-2-(trifluoromethyl)benzene

30913-0250 30913-0050
TS30913-0250EA 249 USD InStock
TS30913-0250 TS30913-0050
4-Chloro-3-(trifluoromethyl)phenyl isocyanate 98%
4-Chloro-3-(trifluoromethyl)phenyl isocyanate


Formula: C₈H₃ClF₃NO
MW: 221.57 g/mol
Boiling Pt: 86…90 °C (14 mmHg)
Melting Pt: 29…37 °C
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00013874
CAS Number: 327-78-6
UN: 2811
ADR: 6.1,I
Order Now
Specification Test Results
Appearance White to yellow Powder and/or chunks or low melting solid
Infrared spectrum Conforms
Melting point 29 to 37 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty