Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Trifluoromethyl)benzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

31296-0050 31296-0250
TS31296-0250EA 326 USD InStock
TS31296-0250 TS31296-0050
3-(Trifluoromethyl)benzylamine 98%
3-(Trifluoromethyl)benzylamine


Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 93…97 °C (22 mmHg)
Density: 1.22 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008117
CAS Number: 2740-83-2
EINECS: 220-367-2
UN: 2735
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4640 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty