Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nordihydroguaiaretic acid 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4,4'-(2,3-Dimethylbutane-1,4-diyl)bis(benzene-1,2-diol)

37630-5000 37630-0050 37630-0010
TS37630-0010EA 101.61 USD InStock
TS37630-0010 TS37630-0050 TS37630-5000
Nordihydroguaiaretic acid 95%
Nordihydroguaiaretic acid


Formula: C₁₈H₂₂O₄
MW: 302.37
Melting Pt: 184…189 °C
Storage Temperature: Freezer
MDL Number: MFCD00002206
CAS Number: 500-38-9
EINECS: 207-903-0
Order Now
Specification Test Results
Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Melting point 184 to 189 °C
HPLC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty