Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-β-Nitrostyrene 97+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (E)-(2-Nitrovinyl)benzenetrans-1-Nitro-2-phenylethylene

38653-0050 38653-1000 38653-0250
TS38653-1000EA 336 USD InStock
TS38653-1000 TS38653-0250 TS38653-0050
trans-β-Nitrostyrene 97+%
trans-β-Nitrostyrene


Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 250…260 °C (760 mmHg)
Melting Pt: 56…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007402
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Yellow Crystals
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty