Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

2-Iodobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: o-Iodobenzonitrile

39180-0250 39180-0050 39180-0010
TS39180-0010EA 30 USD InStock
TS39180-0010 TS39180-0250 TS39180-0050
2-Iodobenzonitrile 99%
2-Iodobenzonitrile


Formula: C₇H₄IN
MW: 229.02 g/mol
Melting Pt: 54 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00079761
CAS Number: 4387-36-4
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Yellow Solid
Infrared spectrum Conforms
Melting point 50 to 54 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty