Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-nitrotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

39842-0250 39842-0050
TS39842-0050EA 41 USD InStock
TS39842-0050 TS39842-0250
2-Bromo-6-nitrotoluene 98%
2-Bromo-6-nitrotoluene


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 143 °C (22 mmHg)
Melting Pt: 38…41 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009792
CAS Number: 55289-35-5
Order Now
Specification Test Results
Appearance Light beige to brown Low melting solid
Infrared spectrum Conforms
Melting point 38 to 42 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty