Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxythiophenol 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: m-Hydroxythiophenol3-Hydroxybenzene-1-thiol3-Hydroxybenzenethiol3-Mercaptophenol3-Hydroxyphenyl mercaptan3-Hydroxythiophenol1-Hydroxy-3-mercaptobenzene

39851-0010 39851-0050
TS39851-0050EA 84 USD InStock
TS39851-0050 TS39851-0010
3-Hydroxythiophenol 96%
3-Hydroxythiophenol


Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 135 °C (35 mmHg)
Melting Pt: 16.5 °C
Density: 1.63 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD01317829
CAS Number: 40248-84-8
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥95 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty