Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: p-Iodobenzoyl chloride

41239-0050 41239-0250
TS41239-0050EA 88 USD InStock
TS41239-0050 TS41239-0250
4-Iodobenzoyl chloride 98%
4-Iodobenzoyl chloride


Formula: C₇H₄ClIO
MW: 266.47 g/mol
Boiling Pt: 100 °C (0.7 mmHg)
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00001058
CAS Number: 1711-02-0
EINECS: 216-974-7
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Appearance Light yellow to beige to light brown Crystalline powder and/or chunks or solid
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥97.5 %
Acidity ≤0.0002 meq/g
Peroxides ≤0.002 % (as H2O2)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty