Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Naphthalenethiol 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Thionaphthol

41527-0050
TS41527-0050EA 172 USD InStock
TS41527-0050
1-Naphthalenethiol 99%
1-Naphthalenethiol


Formula: C₁₀H₈S
Boiling Pt: 131 °C (5 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00039599
CAS Number: 529-36-2
EINECS: 208-462-7
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6790 to 1.6810 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty