Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyloxyacetic acid 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Benzyloxy)acetic acid

42708-0050 42708-0010
TS42708-0050EA 94.28 USD InStock
TS42708-0050 TS42708-0010
Benzyloxyacetic acid 95%
Benzyloxyacetic acid


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 137…139 °C (.6 mmHg)
Density: 1.162 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00274211
CAS Number: 30379-55-6
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
HPLC ≥94 %
NMR (1H-NMR) Consistent with structure

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty