Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6,7-Dimethoxy-1-tetralone 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6,7-Dimethoxy-3,4-dihydronaphthalen-1(2H)-one

44020-0010 44020-0050
TS44020-0050EA 235 USD InStock
TS44020-0050 TS44020-0010
6,7-Dimethoxy-1-tetralone 97%
6,7-Dimethoxy-1-tetralone


Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00134100
CAS Number: 13575-75-2
Order Now
Specification Test Results
Appearance White to off-white to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 95 to 100 °C
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty