Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methyl-5-isoxazolecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Methylisoxazole-5-carboxylic acid

45266-0050
TS45266-0050EA 358 USD InStock
TS45266-0050
3-Methyl-5-isoxazolecarboxylic acid 97%
3-Methyl-5-isoxazolecarboxylic acid


Formula: C₅H₅NO₃
MW: 127.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464222
CAS Number: 4857-42-5
Order Now
Specification Test Results
Appearance Off-white to brown Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty