Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-bromothiazole-4-carboxylate 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethyl 2-bromothiazole-4-carboxylate

45461-0010
TS45461-0010EA 94 USD InStock
TS45461-0010
Ethyl-2-bromothiazole-4-carboxylate 96%
Ethyl-2-bromothiazole-4-carboxylate


Formula: C₆H₆BrNO₂S
MW: 236.08 g/mol
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD03788564
CAS Number: 100367-77-9
Order Now
Specification Test Results
Appearance White to light orange to light brown Powder
Infrared spectrum Conforms
Melting point 66 to 72 °C
GC ≥95.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty