Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Diaminobenzoic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

45749-0010 45749-0050
TS45749-0050EA 159 USD InStock
TS45749-0050 TS45749-0010
2,3-Diaminobenzoic acid 95%
2,3-Diaminobenzoic acid


Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 232…234.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00137818
CAS Number: 603-81-6
Order Now
Specification Test Results
Appearance Off-white to dark brown to black Powder
Infrared spectrum Conforms
HPLC ≥94.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty