Would you like to log in again?
Synergy Transport Tube, SiO₂ Medical Products
Supplier: SiO2 Medical Products
82-9018
76545-530PK 115.49 USD InStock
76545-530
Synergy Transport Tube, SiO₂ Medical Products
Media for Microbiology Prepared Media for Microbiology
The product is a ready-to-use capped and labeled 12 ml tube that is filled with 3 ml of 0.85% saline solution.
  • FDA registered Class I Medical Device
  • Sterile
  • Threaded with red pre-assembled screw caps
  • Leak proof
  • 1-year shelf-life from date of manufacture
A specimen transport and storage container intended to contain biological specimens that may be destroyed or used for diagnostic examination. The container is filled with a solution to preserve the condition of a biological specimen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty