Would you like to log in again?
VWR® Advanced Protection Anti-Skid Shoe Covers
Supplier: VWR
VWR®
SH-X1222-WH SH-X1224-WH SH-X1223-WH
76579-578CS 22.86 USD InStock
76579-578 76579-574 76579-576
VWR® Advanced Protection Anti-Skid Shoe Covers
Disposable Garments
VWR® Advanced protection anti-skid shoe covers with elastic band on the front and bottom, made of durable 49 g laminated fabric.
  • Elastic band on front and bottom
  • Made of durable 49 g laminated fabric
  • Multiple sizes available
  • White finish
Shoecovers are supplied in a case of 150 pairs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty