Would you like to log in again?
VWR® C-Leg Lab Benches, Frame + Top + Uprights + Shelves, Manual, Snow White
Supplier: Avantor
VWR®
14-C100SW429 14-C100SW420 14-C100SW426 14-C100SW431 14-C100SW433 14-C100SW439 14-C100SW423 14-C100SW428 14-C100SW430 14-C100SW436 14-C100SW427 14-C100SW421 14-C100SW435 14-C100SW424 14-C100SW425 14-C100SW434 14-C100SW422 14-C100SW432 14-C100SW437 14-C100SW438
76551-150EA 2737.81 USD InStock
76551-150 76550-804 76550-518 76551-124 76550-544 76550-566 76550-456 76550-788 76550-448 76550-954 76550-866 76550-734 76550-966 76550-428 76550-370 76550-294 76550-792 76550-694 76550-652 76550-740
VWR® C-Leg Lab Benches, Frame + Top + Uprights + Shelves, Manual, Snow White
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel. There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality.
  • Snow white frame
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Offering a wide range of accessories to meet any need
  • Various concept workbench sizes and combinations
  • Height adjustment available either electrically (electric motor height adjustment) or with a hand crank or an Allen key
  • A wide range of accessories and add-ons, making concept workbenches easy to customize
  • Impressive stability with its steel frame and maximum load capacity of 1100 lbs.
  • Cleanroom suitability ISO class 7
The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Shelves depth 11.81".
Accessories information: Upright accessories can be added in any position along the upright in 1" increments. Some accessories are bench style or size dependent.
Ordering information: Manual, hand crank, and motorized frame styles are available, as well as several configurations, including frame only, frame + top, frame + top + uprights, frame + top + uprights + shelves, and frames with notches for uprights (all sold separately). Contact your Avantor representative for more information.
Notched work surfaces add 2.625" to depth of table frame. Work surfaces for single bay uprights include two notches (left and right); work surfaces for double bay uprights include three notches (left, center, right).
Tables ship LTL, flat-packed. Installation not included.
Packaging: Benches are shipped knocked down, on non-stackable pallets. Assembly instructions and hardware included.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty